Skip to Content
Merck
All Photos(2)

Key Documents

756792

Sigma-Aldrich

3,7-Diacetyl-1,3,7-triaza-5-phosphabicyclo[3.3.1]nonane

97%

Synonym(s):

DAPTA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16N3O2P
CAS Number:
Molecular Weight:
229.22
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

solid

mp

183 °C (decomposition)

functional group

phosphine

SMILES string

CC(=O)N1CN2CN(CP(C2)C1)C(C)=O

InChI

1S/C9H16N3O2P/c1-8(13)11-3-10-4-12(9(2)14)7-15(5-10)6-11/h3-7H2,1-2H3

InChI key

MYJHMDGWZWBLCK-UHFFFAOYSA-N

Application

3,7-Diacetyl-1,3,7-triaza-5-phosphabicyclo[3.3.1]nonane is a phosphine ligand and acetylized derivative of 1,3,5-triaza-7-phosphaadamantane[1][2] that can be used:
  • As a ligand in the synthesis of diiron dithiolate complexes of Fe-only hydrogenase to improve the hydro- and protophilicity of the active site.[2]
  • To prepare (η6-arene)-ruthenium(II) complex, which is employed as a catalyst in the reduction of allylic alcohols.[3]
  • As a ligand in the synthesis of water-soluble cis- and trans-Pt(II) and Pd(II) metal complexes.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruthenium-catalyzed reduction of allylic alcohols using glycerol as solvent and hydrogen donor
Diaz-Alvarez AE, et al.
Catalysis Communications, 13(1), 91-96 (2011)
Yanfang Song et al.
Cancer research, 77(7), 1611-1622 (2017-01-14)
Mice housed in an enriched environment display a tumor-resistant phenotype due to eustress stimulation. However, the mechanisms underlying enriched environment-induced protection against cancers remain largely unexplained. In this study, we observed a significant antitumor effect induced by enriched environment in
Synthesis and characterization of Pt(II) and Pd(II) PTA and DAPTA complexes
Braddock-Wilking J, et al.
Polyhedron, 79(1), 16-28 (2014)
Chen Chen et al.
Cellular and molecular neurobiology, 39(5), 651-669 (2019-04-15)
CKLF1 is a chemokine with increased expression in ischemic brain, and targeting CKLF1 has shown therapeutic effects in cerebral ischemia model. Microglia/macrophage polarization is a mechanism involved in poststroke injury expansion. Considering the quick and obvious response of CKLF1 and
An approach to water-soluble hydrogenase active site models: Synthesis and electrochemistry of diiron dithiolate complexes with 3, 7-diacetyl-1, 3, 7-triaza-5-phosphabicyclo [3.3. 1] nonane ligand (s)
Na Y, et al.
Journal of Organometallic Chemistry, 691(23), 5045-5051 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service