755303
3-Amino-2-fluoropropionic acid
95%
Synonym(s):
DL-2-Fluoro-β-alanine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
95%
form
solid
mp
240-245 °C
functional group
amine
carboxylic acid
fluoro
SMILES string
NCC(F)C(O)=O
InChI
1S/C3H6FNO2/c4-2(1-5)3(6)7/h2H,1,5H2,(H,6,7)
InChI key
OJQNRNQELNLWHH-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
American journal of ophthalmology, 142(1), 194-196 (2006-07-04)
The aim of this study is to determine if systemic absorption is present after 5-fluorouracil (5FU) is used in the vitrectomy infusion. A two-case clinical study. A catabolite of 5FU, alpha-fluoro-beta-alanine (FBAL), was measured in urine samples of two patients
Biomedical chromatography : BMC, 20(3), 257-266 (2005-09-17)
The validation of an analytical method for the measurement of the unnatural amino acid alpha-fluoro-beta-alanine (AFBA), the main metabolite of the antineoplastic drug 5-fluorouracil (5FU), in urine for the biological monitoring of the exposure of hospital workers to the drug
The American journal of physiology, 276(1 Pt 1), G206-G210 (1999-01-14)
Studies on the compartmentalization of uridine catabolic metabolism in liver have indicated accumulation of beta-alanine as well as alpha-fluoro-beta-alanine (FbetaAL) for 5-fluorouracil in the hepatocytes. Using preparations of rat hepatocytes we were able to identify a Na+-dependent transport with high
Magnetic resonance in medicine, 50(2), 303-308 (2003-07-24)
Fluorine MR spectroscopy ((19)F MRS) is an indispensable tool for assessing the pharmacokinetics of fluorinated drugs. Since the metabolism of 5-fluorouracil (5FU), a frequently used cytotoxic drug, is expected to be different in normal liver and in tumor tissue, spatial
Magnetic resonance in medicine, 37(3), 321-326 (1997-03-01)
An RF network and a dual-tuned surface coil are described for obtaining proton-decoupled, NOE enhanced 19F spectra from a whole body clinical imager operating at 1.5 Tesia. The network removes 19F frequency noise from the decoupler transmitter, and prevents preamplifier
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service