Skip to Content
Merck
All Photos(1)

Key Documents

750050

Sigma-Aldrich

N-(4-Formylphenyl)carbazole

97%

Synonym(s):

4-(9H-Carbazol-9-yl)benzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H13NO
CAS Number:
Molecular Weight:
271.31
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

155-160 °C

SMILES string

O=Cc1ccc(cc1)-n2c3ccccc3c4ccccc24

InChI

1S/C19H13NO/c21-13-14-9-11-15(12-10-14)20-18-7-3-1-5-16(18)17-6-2-4-8-19(17)20/h1-13H

InChI key

RPHLDCKUUAGNAC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service