Skip to Content
Merck
All Photos(2)

Key Documents

74988

Sigma-Aldrich

Octylamine

≥99.5%

Synonym(s):

1-Aminooctane, n-Octylamine, Caprylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7NH2
CAS Number:
Molecular Weight:
129.24
Beilstein:
1679227
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

impurities

≤0.2% water

refractive index

n20/D 1.429 (lit.)
n20/D 1.429

bp

175-177 °C (lit.)

mp

−5-−1 °C (lit.)

transition temp

solidification point −1-1 °C

density

0.782 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCN

InChI

1S/C8H19N/c1-2-3-4-5-6-7-8-9/h2-9H2,1H3

InChI key

IOQPZZOEVPZRBK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Octylamine can be used as:
  • A surfactant in the synthesis of mesoporous nickel hydroxy nitrates for fuel cell applications.
  • A structure-directing agent to prepare sandwich-type ZnS/octylamine nanosheets, applicable in the fabrication of optical and electronic devices.
  • An organic fuel in the reduction of ferric nitrate to magnetic particles having a high magnetic moment.
  • A solvent and intercalation agent in the synthesis of ultra-expanded carbon-containing vanadium disulfide composite materials.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Engineering ultra-enlarged interlayer carbon-containing vanadium disulfide composite for high-performance sodium and potassium ion storage
Xie X-C, et al.
Journal of alloys and compounds, 847(9), 156288-156288 (2020)
Synthesis of bimodal mesoporous Ni oxide from octylamine
Feng F, et al.
Journal of Sol-Gel Science and Technology, 51(2), 153-157 (2009)
Hydrothermal synthesis of novel sandwich-like structured ZnS/octylamine hybrid nanosheets
Li J, et al.
Solid State Communications, 130(9), 619-622 (2004)
Octylamine as a novel fuel for the preparation of magnetic iron oxide particles by an aqueous auto-ignition method
Ben-Arfa BAE, et al.
Journal of alloys and compounds, 805(9), 545-550 (2019)
Rosario Zamora et al.
Journal of agricultural and food chemistry, 54(6), 2398-2404 (2006-03-16)
The reactions of 4,5-epoxy-2-decenal with octylamine, benzylamine, and 2-phenylglycine methyl ester were studied to investigate if amines may suffer a Strecker type degradation by epoxyalkenals analogously to amino acids. In addition to other reactions, the studied amines were converted into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service