747181
Poly(ethylene glycol)-block-poly(N-isopropylacrylamide)
α-methoxy, ω-dodecane, PEG Mn 2,000, PNIPAM Mn 24,000, PDI <1.2
Synonym(s):
PEG-block-PNIPAM, PEG-b-polyNIPAM, PEGylated polyNIPAM, PNIPAAm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(OC2H4)mC6H7NO2(C6H11NO)nH
UNSPSC Code:
12352100
NACRES:
NA.23
Recommended Products
Related Categories
General description
Poly(ethylene glycol)-b-poly(N-isopropylacrylamide) (PEG110-b-PNIPAM44) undergoes thermoresponsive micellization in water, forming micelles of varied size, structure and distribution depending on the concentration of the block copolymer.
Application
Well-defined PEG-block-PNIPAM diblock copolymers can form thermoresponsive vesicles in water and enable controlled drug release.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Thermoresponsive micellization of poly (ethylene glycol)-b-poly (N-isopropylacrylamide) in water.
Zhang W
Macromolecules, 38(13), 5743-5747 (2005)
Temperature-Controlled Assembly and Release from Polymer Vesicles of Poly(ethylene oxide)-block-poly(N-isopropylacrylamide)
Qin S, et al.
Advanced Materials, 18, 2905?2909-2905?2909 (2006)
Articles
Professor Mitsuhiro Ebara provides insights on several types of smart nanofiber mesh systems that have been explored for different drug delivery purposes.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service