Skip to Content
Merck
All Photos(2)

Documents

747130

Sigma-Aldrich

Josiphos SL-J009-1 Pd G3

Synonym(s):

Josiphos SL-J009-1-G3-palladacycle, {(R)-1-[(Sp)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldi-tert-butylphosphine}[2-(2′-amino-1,1′-biphenyl)]palladium(II) methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H65FeNO3P2PdS
CAS Number:
Molecular Weight:
924.28
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

feature

generation 3

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

180-186 °C

functional group

phosphine

SMILES string

NC1=C(C=CC=C1)C2=C([Pd]OS(C)(=O)=O)C=CC=C2.C[C@@H](P(C(C)(C)C)C(C)(C)C)[C]3[C](P(C4CCCCC4)C5CCCCC5)[C][C][C]3.[C]6[C][C][C][C]6.[Fe]

InChI

1S/C27H47P2.C12H10N.C5H5.CH4O3S.Fe.Pd/c1-21(29(26(2,3)4)27(5,6)7)24-19-14-20-25(24)28(22-15-10-8-11-16-22)23-17-12-9-13-18-23;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-2-4-5-3-1;1-5(2,3)4;;/h14,19-23H,8-13,15-18H2,1-7H3;1-6,8-9H,13H2;1-5H;1H3,(H,2,3,4);;/q;;;;;+1/p-1/t21-;;;;;/m1...../s1

InChI key

GJFJVCUFUWXFEB-RPDSBUJNSA-M

General description

Josiphos SL-J009-1 Pd G3 (Josiphos SL-J009-1-G3-palladacycle) is a modified G3 precatalyst (G3′). It has been synthesized by Buchwald and coworkers from second generation (G2) precatalyst by methylation of the amino group present on biphenyl backbone. It is a useful catalyst for various cross-coupling reactions. G3 precatalysts are versatile catalysts. They have long life in solution and are readily soluble in various organic solvents. G3- precatalysts have been employed in various C-N bond forming reactions.

Application


  • Total Syntheses of N-Paspaline and N-Emindole PB: Discusses the use of various catalysts including RuPhos and Josiphos ligands for synthesizing complex molecular structures (Radical-Polar Crossover Cyclizations, 2020).

  • Unprotected Indazoles Are Resilient to Ring-Opening Isomerization: A Case Study on Catalytic C-S Couplings in the Presence of Strong Base: Utilizes Josiphos SL-J009-1 Pd-G3 for catalytic C-S coupling reactions under strong base conditions (The Journal of Organic Chemistry, 2017).

  • Synthesis and characterization of an isopropylBippyPhos precatalyst: Compares various phosphine ligands including XPhos and Josiphos for catalytic activity in organic syntheses (Tetrahedron, 2022).

  • Novel quorum sensing inhibitors targeting PqsR: Mentions the experimental use of Josiphos ligands in the synthesis of potential pharmaceutical compounds (2020, University of Saarland).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-Aminobiphenyl Palladacycles: The ?Most Powerful? Precatalysts in C-C and C-Heteroatom Cross-Couplings.
Bruneau A, et al.
ACS Catalysis, 5(2), 1386-1396 (2015)

Articles

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

See All

Related Content

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service