Skip to Content
Merck
All Photos(1)

Key Documents

744050

Sigma-Aldrich

(Difluoromethyl)trimethylsilane

≥98.0% (GC)

Synonym(s):

(Trimethylsilyl)difluoromethane, Trimethyl(difluoromethyl)silane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCHF2
CAS Number:
Molecular Weight:
124.20
Beilstein:
2423974
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

liquid

functional group

fluoro

SMILES string

C[Si](C)(C)C(F)F

InChI

1S/C4H10F2Si/c1-7(2,3)4(5)6/h4H,1-3H3

InChI key

OOKFLLNDYNWCHK-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc Tordeux et al.
Magnetic resonance in chemistry : MRC, 42(8), 700-703 (2004-07-13)
Isotope effects on fluorine chemical shifts induced by heteroatoms bonded covalently to a carbon atom bearing fluorine atoms were studied. For each compound, the isotope-induced chemical shifts 2delta19F(X) through two bonds were measured for the heteroatom (X = 29/28Si, 30/28Si
Yanchuan Zhao et al.
Organic letters, 13(19), 5342-5345 (2011-09-14)
Since 1995, Me(3)SiCF(2)H has been widely believed to be an inefficient difluoromethylating agent, which requires harsh reaction conditions to cleave its rather inert Si-CF(2)H bond. However, it has now been found that, by using a proper Lewis base activator, Me(3)SiCF(2)H

Related Content

Prof. Jinbo Hu's lab focuses on developing new fluorination reagents and reactions, including difluoromethylation and monofluoromethylation.

Prof. Jinbo Hu's lab focuses on developing new fluorination reagents and reactions, including difluoromethylation and monofluoromethylation.

Prof. Jinbo Hu's lab focuses on developing new fluorination reagents and reactions, including difluoromethylation and monofluoromethylation.

Prof. Jinbo Hu's lab focuses on developing new fluorination reagents and reactions, including difluoromethylation and monofluoromethylation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service