Skip to Content
Merck
All Photos(1)

Key Documents

735817

Sigma-Aldrich

Polypyrrole-block-poly(caprolactone)

0.3-0.7 wt. % (dispersion in nitromethane), contains p-toluenesulfonate as dopant

Synonym(s):

Biotron PP-NM, PCL-block-PPy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

form

liquid (dispersion)

Quality Level

contains

p-toluenesulfonate as dopant

composition

Biotron PP polymer, 0.3-0.7 wt. %
ethanol, 4-8 wt. %
nitromethane, 90-95 wt. %

concentration

0.3-0.7 wt. % (dispersion in nitromethane)

conductivity

10-40 S/cm (bulk)

General description

Sheet resistance, spin cast thin films: 10,000-50,000 Ohms/sq

Average transmittance: 70-90%

Biodegradble conducting polymer for bomedical applications. PCL diol blocks have average molecular weight 2000 and PPy block have an average molecular weight of 4000 with a 25% doping level. Good biocompatibility with expected weight loss of 40% at 200 days in pH=7 buffer.

Application

Thin films can be prepared by spin casting the nitromethane dispersion at 1000 rpm. To spin coat a thin film at 1000 RPM, 30 seconds to 1 minute is usually enough time to spread this dispersion. Thicker and more conducting films can be prepared by air spraying the dispersion on a substrate. Any substrate that is compatible can be used. Sprayed films can be dried at 30-40 °C, then pressed between two polished dye plates at 1000-10,000 psi to increase film density, strength and scratch resistance. The drying time of the air sprayed film depends on the thickness of the prepared film. Try weighing the sample periodically during drying.

Features and Benefits

The block copolymer structure renders the conducting polymer polypyrrole dispersible in organic solvents and biodegradable.

Caution

Store this product at room temperature (do not refrigerate or freeze). Severe settling and separation will normally occur. Agitate, sonicate, and filter prior to use. A plug of glass wool in a pipette can be used for easy filtration. The shelf life of this product will vary depending on how it is stored and handled. Product should be discarded if heavy aggregation occurs, or if sonication / filtering are inadequate to produce a good casting solution. When spray coating is used, the user should take every precaution necessarily to prevent exposure to solvent vapors from this product

Other Notes

Will settle over time. Must be sonicated before use

Legal Information

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

66.9 °F

Flash Point(C)

19.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Materials for Bioelectronic and
Biomedical Applications
Zhang Y, et al.
Material Matters, 8(1) null
Chemical Synthesis and Morphology of b?Naphthalene Sulfonic Acid-Doped Polypyrrole Micro/Nanotube.
Akinyeye R, et al.
Fullerenes, Nanotubes, and Carbon Nanostructures, 14 (2006)
Synergic effect in electrical conductivity using a combination of two fillers in PVDF hybrids composites.
da Silva AB, et al.
European Polymer Journal, 49(10), 3318-3327 (2013)
Polypropylene composites for polymer electrolyte membrane fuel cell bipolar plates.
Yeetsorn R, et al.
Macromolecular Symposia, 264(1) (2008)
Soluble conducting polypyrrole doped with DBSA?CSA mixed acid
Shen Y and Wan M
Journal of Applied Polymer Science, 68(8), 1277-1284 (1998)

Articles

The application of conducting polymers at the interface with biology is an exciting new trend in organic electronics research.

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service