Skip to Content
Merck
All Photos(1)

Key Documents

733059

Sigma-Aldrich

Ethyl 3-hydroxycyclohexanecarboxylate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O3
CAS Number:
Molecular Weight:
172.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

density

1.067 g/mL at 25 °C

SMILES string

CCOC(=O)C1CCCC(O)C1

InChI

1S/C9H16O3/c1-2-12-9(11)7-4-3-5-8(10)6-7/h7-8,10H,2-6H2,1H3

InChI key

BGAOLPQGOBDXBH-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service