Skip to Content
Merck
All Photos(1)

Key Documents

729698

Sigma-Aldrich

(S)-(+)-Pantolactone

97%

Synonym(s):

(S)-(+)-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, (S)-(+)-β,β-Dimethyl-α-hydroxy-γ-butyrolactone, (S)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, L-Pantolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
Beilstein:
4659425
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

crystals

optical activity

[α]22/D +50.0°, c = 0.5 in H2O

bp

120-122 °C/15 mmHg (lit.)

mp

89-93 °C

functional group

ester
hydroxyl

storage temp.

2-8°C

SMILES string

CC1(C)COC(=O)[C@H]1O

InChI

1S/C6H10O3/c1-6(2)3-9-5(8)4(6)7/h4,7H,3H2,1-2H3/t4-/m1/s1

InChI key

SERHXTVXHNVDKA-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiqiang Liu et al.
Journal of agricultural and food chemistry, 54(16), 5823-5830 (2006-08-03)
D-Pantonohydrolase has attracted increasing attention as a biocatalyst for stereospecific production of D-pantoic acid. The Fusarium moniliforme D-pantonohydrolase was selected for directed evolution through error-prone Polymerase Chain Reaction (PCR) combined with DNA shuffling for improved activity and pH stability using
Michael-Rock Goldsmith et al.
Journal of the American Chemical Society, 125(51), 15696-15697 (2003-12-18)
Dilute solutions of (R)-(-)-pantolactone in CCl4 were studied by polarimetry in conjunction with theoretical calculations of [alpha]D. Our data demonstrate that the self-association of a chiral solute results in a change in [alpha]D that can be accounted for by the
Total synthesis and antitumor activity of ZK-EPO: the first fully synthetic epothilone in clinical development.
Ulrich Klar et al.
Angewandte Chemie (International ed. in English), 45(47), 7942-7948 (2006-09-29)
A D Keefe et al.
Nature, 373(6516), 683-685 (1995-02-23)
The involvement of coenzyme A in many enzyme reactions suggests that it acted in this capacity very early in the development of life on Earth. Particularly relevant in this regard is its role in the activation of amino acids and
M Kataoka et al.
Applied microbiology and biotechnology, 64(3), 359-366 (2003-11-01)
The genes encoding two conjugated polyketone reductases (CPR-C1, CPR-C2) of Candida parapsilosis IFO 0708 were cloned and sequenced. The genes encoded a total of 304 and 307 amino acid residues for CPR-C1 and CPR-C2, respectively. The deduced amino acid sequences

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service