Skip to Content
Merck
All Photos(3)

Key Documents

729221

Sigma-Aldrich

trans-2-(Pinacol boronate)vinylboronic acid MIDA ester

96%

Synonym(s):

(E)-(2-Pinacolethenyl)boronate MIDA ester, trans-(2-Pinacolethenyl)boronate MIDA ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H21B2NO6
Molecular Weight:
308.93
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

mp

254-260 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)\C=C\B2OC(C)(C)C(C)(C)O2

InChI

1S/C13H21B2NO6/c1-12(2)13(3,4)22-15(21-12)7-6-14-19-10(17)8-16(5)9-11(18)20-14/h6-7H,8-9H2,1-5H3/b7-6+

InChI key

MVTNIQCFWSPASI-VOTSOKGWSA-N

Legal Information

Patents pending. U.S. Patent App Ser. Nos. 60/951,405 and 11/937,338 and Internat. App No. PCT/US07/84156 may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service