728950
2-Bromopropionaldehyde diethyl acetal
95%
Synonym(s):
2-Bromo-1,1-diethoxypropane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H15BrO2
CAS Number:
Molecular Weight:
211.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.439
density
1.198 g/mL at 25 °C
functional group
bromo
ether
SMILES string
CCOC(OCC)C(C)Br
InChI
1S/C7H15BrO2/c1-4-9-7(6(3)8)10-5-2/h6-7H,4-5H2,1-3H3
InChI key
AHIUAFXWZKCJLR-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
145.0 °F
Flash Point(C)
62.8 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service