Skip to Content
Merck
All Photos(1)

Key Documents

726060

Sigma-Aldrich

5-(Bromomethyl)-1,3-benzodioxole

96%

Synonym(s):

3,4-Methylenedioxybenzyl bromide, Piperonyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7BrO2
CAS Number:
Molecular Weight:
215.04
Beilstein:
135376
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.5% (GC)
95.5-104.5% (T)
96%

form

crystals

storage temp.

−20°C

SMILES string

BrCc1ccc2OCOc2c1

InChI

1S/C8H7BrO2/c9-4-6-1-2-7-8(3-6)11-5-10-7/h1-3H,4-5H2

InChI key

UNYHRXLMTSXVIB-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service