Skip to Content
Merck
All Photos(1)

Key Documents

725765

Sigma-Aldrich

2-Methyl-4-isothiazolin-3-one

95%

Synonym(s):

2-Methyl-3(2H)-isothiazolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5NOS
CAS Number:
Molecular Weight:
115.15
Beilstein:
606203
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Assay

≥94.0% (GC)
95%

form

lumps

impurities

≤5% water

SMILES string

CN1SC=CC1=O

InChI

1S/C4H5NOS/c1-5-4(6)2-3-7-5/h2-3H,1H3

InChI key

BEGLCMHJXHIJLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Insights into the mechanisms of within-species variation in sensitivity to chemicals: A case study using daphnids exposed to CMIT/MIT biocide.: This study investigates the differential sensitivity of daphnids to 2-Methyl-4-isothiazolin-3-one, highlighting the biochemical mechanisms underlying these variations. The findings provide crucial insights for environmental safety assessments (Kim et al., 2023).
  • Effects of the biocide methylisothiazolinone on Xenopus laevis wound healing and tail regeneration.: The study assesses the impact of 2-Methyl-4-isothiazolin-3-one on regenerative processes in amphibians, contributing to the understanding of its biological effects and potential toxicity (Delos Santos et al., 2016).
  • T cell lymphomatoid contact dermatitis: a challenging case and review of the literature.: This review includes discussions on 2-Methyl-4-isothiazolin-3-one-induced contact dermatitis, providing valuable information for dermatological applications and safety assessments (Knackstedt and Zug, 2015).

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1A

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leena Ackermann et al.
Contact dermatitis, 64(1), 49-53 (2010-12-21)
Antimicrobials constitute the second most common cause of contact allergy to cosmetics. Methylisothiazolinone (MI), previously always used together with methylchloroisothiazolinone (MCI), has recently been approved in the EU for use on its own in cosmetics and also various industrial products.
Michael D Lundov et al.
Contact dermatitis, 69(5), 271-275 (2013-10-15)
The prevalence of contact allergy to the isothiazolinone preservative methylchloroisothiazolinone (MCI) in combination with methylisothiazolinone (MI) and MI alone has increased in the last couple of years. To investigate the prevalence of contact allergy to MI, MCI/MI and benzisothiazolinone (BIT)
Julien Mutschler et al.
Toxicology in vitro : an international journal published in association with BIBRA, 23(3), 439-446 (2009-05-16)
Assessment of skin sensitization hazard of chemicals currently depends on in vivo methods. Considering the forthcoming European Union ban on in vivo testing of cosmetic/toiletry ingredients, the search for alternative non-animal approaches is an urgent challenge for investigators today. For
Eleanor Higgins et al.
Dermatitis : contact, atopic, occupational, drug, 24(2), 73-76 (2013-03-12)
The isothiazolinones methylchloroisothiazolinone and methylisothiazolinone (MCI/MI) are the active ingredients in a frequently used preservative in cosmetic, household, and industrial products. This study reviewed our department's cases of allergic contact dermatitis caused by MCI/MI, outlining their clinical presentation and possible
A Spawn et al.
Neuroscience, 205, 194-204 (2012-01-17)
Methylisothiazolinone (MIT) is a commonly used biocide known to be neurotoxic in vitro. Brief exposure of cortical neurons in culture to MIT results in increased neurodegeneration, whereas chronic exposure of developing neurons in culture to low concentrations of MIT has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service