Skip to Content
Merck
All Photos(2)

Documents

719951

Sigma-Aldrich

Resomer® R 202 S, Poly(D,L-lactide)

ester terminated, viscosity 0.16-0.24 dL/g 

Synonym(s):

Lactide polymer, PDLLA, Polylactide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H8O4)n
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

form

amorphous

degradation timeframe

<6 months

viscosity

0.16-0.24 dL/g

transition temp

Tg 38-42 °C

storage temp.

2-8°C

Application

Controlled release

Legal Information

Product of Evonik
RESOMER is a registered trademark of Evonik Rohm GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Interest in utilizing biodegradable polymers for biomedical applications has grown since the 1960s.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service