Skip to Content
Merck
All Photos(1)

Documents

709972

Sigma-Aldrich

N-Boc-2,3-dihydro-1H-pyrrole

95%

Synonym(s):

1-(tert-Butoxycarbonyl)-2,3-dihydro-1H-pyrrole, N-Boc-2-pyrroline, tert-Butyl 2,3-dihydro-1H-pyrrole-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NO2
CAS Number:
Molecular Weight:
169.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.468

density

0.980 g/mL at 25 °C

SMILES string

CC(C)(C)OC(=O)N1CCC=C1

InChI

1S/C9H15NO2/c1-9(2,3)12-8(11)10-6-4-5-7-10/h4,6H,5,7H2,1-3H3

InChI key

DCVICHWBECIALB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service