706094
6-Hydroxypyridine-3-boronic acid pinacol ester
97%
Synonym(s):
2-Hydroxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Hydroxypyridine-5-boronic acid, pinacol ester
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C11H16BNO3
CAS Number:
Molecular Weight:
221.06
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
196-200 °C
SMILES string
CC1(C)OB(OC1(C)C)c2ccc(O)nc2
InChI
1S/C11H16BNO3/c1-10(2)11(3,4)16-12(15-10)8-5-6-9(14)13-7-8/h5-7H,1-4H3,(H,13,14)
InChI key
WAUWXCUPDOXYKS-UHFFFAOYSA-N
Application
6-Hydroxypyridine-3-boronic acid pinacol ester can be used as a reactant:
- In the synthesis of novel glutamate transporter EAAT2 activators.
- To prepare crizotinib derivatives as SHIP2 inhibitors.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Structure-activity relationship study of pyridazine derivatives as glutamate transporter EAAT2 activators
Xing X, et al.
Bioorganic & Medicinal Chemistry Letters, 21(19), 5774-5777 (2011)
Identification of crizotinib derivatives as potent SHIP2 inhibitors for the treatment of Alzheimer's disease
Lim JW, et al.
European Journal of Medicinal Chemistry, 157(19), 405-422 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service