Skip to Content
Merck
All Photos(3)

Documents

702668

Sigma-Aldrich

Thieno[3,2-b]thiophene

95%

Synonym(s):

1,4-Dithiapentalene, 1,4-Thiophthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4S2
CAS Number:
Molecular Weight:
140.23
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

mp

53-58 °C

SMILES string

c1cc2sccc2s1

InChI

1S/C6H4S2/c1-3-7-6-2-4-8-5(1)6/h1-4H

InChI key

VJYJJHQEVLEOFL-UHFFFAOYSA-N

General description

Thieno(3,2-b)thiophene (TT) is an electron rich conjugated polymer that has a quinoidal structure with a narrow band gap. It facilitates a strong intermolecular networking.

Application

TT may be used as conjugated side chains in the synthesis of donor-acceptor copolymers. It finds potential applications in the enhancement of power conversion efficiency (PCE) for organic solar cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

210.0 °F

Flash Point(C)

98.9 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrei Chernyshev et al.
Polymers, 13(3) (2021-01-28)
Four new bis(tpy) unimers with different linkers between the thieno[3,2-b]thiophene-2,5-diyl central unit and terpyridine-4'-yl (tpy) end-groups: no linker (Tt), ethynediyl (TtE), 1,4-phenylene (TtPh) and 2,2'-bithophene-5,5'-diyl (TtB) are prepared, characterized, and assembled with Fe2+ ions to metallo-supramolecular polymers (Fe-MSPs). The Fe-MSP
Synthesis and photovoltaic properties of 2D-conjugated polymers with alkylsilyl-substituted thieno (3, 2-b) thiophene conjugated side chains
Yan T, et al.
Organic Electronics, 57(41), 255-262 (2018)
A solution-processed high performance organic solar cell using a small molecule with the thieno (3, 2-b) thiophene central unit
Zhang, Q, et al.
Chemical Communications (Cambridge, England), 51(83), 15268-15271 (2015)
A fused thieno (3, 2-b) thiophene-dithiophene based donor molecule for organic photovoltaics: a structural comparative study with indacenodithiophene
Abe Y, et al.
Journal of Material Chemistry C, 4(41), 9656-9663 (2016)
Hocheon Yoo et al.
Nature communications, 10(1), 2424-2424 (2019-06-05)
Multilevel metal interconnects are crucial for the development of large-scale organic integrated circuits. In particular, three-dimensional integrated circuits require a large number of vertical interconnects between layers. Here, we present a novel multilevel metal interconnect scheme that involves solvent-free patterning

Articles

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service