Skip to Content
Merck
All Photos(2)

Documents

69822

Sigma-Aldrich

Methyl DL-lactate

greener alternative

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)COOCH3
CAS Number:
Molecular Weight:
104.10
Beilstein:
1720586
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

liquid

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.413

density

1.093 g/mL at 20 °C (lit.)

greener alternative category

SMILES string

COC(=O)C(C)O

InChI

1S/C4H8O3/c1-3(5)4(6)7-2/h3,5H,1-2H3

InChI key

LPEKGGXMPWTOCB-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry.This compound has benign qualities and is biodegradable, water miscible, and functions as a versatile solvent for CA membrane preparation. This is also used as an intermediate for the production of other chemicals, polymers, and derivatives. Thus this product has been enhanced for Safer solvents and auxiliaries. Click here for more information.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chirped-pulse and cavity-based Fourier transform microwave spectra of the methyl lactate···ammonia adduct.
Javix Thomas et al.
Angewandte Chemie (International ed. in English), 52(16), 4402-4405 (2013-03-16)
A Knüttel et al.
Magnetic resonance in medicine, 9(2), 254-260 (1989-02-01)
An RF and field gradient pulse sequence is presented permitting the single-scan volume-selective spectral editing of the signals of coupled spins. The attribute "single scan" means that the editing of resonance lines is performed in each accumulation or phase cycling
Opposite signs of capacitive microsensor signals upon exposure to the enantiomers of methyl propionate compounds.
Petra Kurzawski et al.
Angewandte Chemie (International ed. in English), 47(5), 913-916 (2007-12-22)
K Le Barbu-Debus et al.
Physical chemistry chemical physics : PCCP, 11(35), 7589-7598 (2009-12-03)
Complexation between (1R,2S)-(+)-cis-1-amino-2-indanol (AI) and the two enantiomers of methyl lactate has been studied by means of laser-induced fluorescence, resonance-enhanced two-photon ionisation, and IR-UV double resonance spectroscopy, in the region of 3 microm. Two isomeric complexes have been spectroscopically characterised
Nicole Borho et al.
Organic & biomolecular chemistry, 1(23), 4351-4358 (2003-12-20)
Chiral recognition and subsequent selective self-organisation into hydrogen-bonded n-mers is observed in supersonic methyl lactate expansions. The nu(OH) and nu(C=O)-vibrations are investigated by ragout-jet FTIR-spectroscopy and lead to the assignment of homo- and heterochiral clusters of at least three different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service