697931
N-Hydroxyethyl acrylamide
contains 1,000 ppm monomethyl ether hydroquinone as stabilizer, 97%
Synonym(s):
N-(2-Hydroxyethyl)acrylamide, HEAA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
97%
form
liquid
contains
1,000 ppm monomethyl ether hydroquinone as stabilizer
refractive index
n20/D 1.505
bp
129-130 °C
density
1.111 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
OCCNC(=O)C=C
InChI
1S/C5H9NO2/c1-2-5(8)6-3-4-7/h2,7H,1,3-4H2,(H,6,8)
InChI key
UUORTJUPDJJXST-UHFFFAOYSA-N
Related Categories
Application
N-Hydroxyethyl acrylamide is an amphipathic polymer that can be used for a variety of applications such as:
- preparation of hydrogels for the formation of a stimulo-responsive polymer for cell signaling and bioengineering research
- in the synthesis of lanthanide based luminescent materials for sensors and bio-imaging
- formation of adhesive hydrogel for potential usage in the smart adhesives
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - STOT RE 2 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
298.4 °F - closed cup
Flash Point(C)
148 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Urara Hasegawa et al.
Macromolecular bioscience, 16(7), 1009-1018 (2016-03-10)
Furoxans, or 1,2,5-oxadiazole-N-oxides, are a class of nitric oxide (NO)-donating compounds that release NO in response to thiol-containing molecules. In this study, polymeric micelles bearing furoxan moieties are prepared from an amphiphilic block copolymer consisting of a hydrophobic furoxan-bearing block
Sifani Zavahir et al.
Nanomaterials (Basel, Switzerland), 10(7) (2020-07-28)
Soft actuators based on hydrogel materials, which can convert light energy directly into mechanical energy, are of the utmost importance, especially with enhancements in device development. However, the hunt for specific photothermal nanomaterials with distinct performance remains challenging. In this
Xinyu Hu et al.
ChemMedChem, 12(19), 1600-1609 (2017-09-01)
Salecan is a water-soluble extracellular β-glucan and has excellent physicochemical and biological properties for hydrogel preparation. In this study, a new pH/magnetic field dual-responsive hydrogel was prepared by the graft copolymerization of salecan with 4-pentenoic acid (PA) and N-hydroxyethylacrylamide (HEAA)
pH responsive and oxidation resistant wet adhesive based on reversible catechol-boronate complexation
Narkar AR, et al.
Chemistry of Materials, 28(15), 5432-5439 (2016)
Mai N Vu et al.
Small (Weinheim an der Bergstrasse, Germany), 16(33), e2002861-e2002861 (2020-06-26)
A key concept in nanomedicine is encapsulating therapeutic or diagnostic agents inside nanoparticles to prolong blood circulation time and to enhance interactions with targeted cells. During circulation and depending on the selected application (e.g., cancer drug delivery or immune modulators)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service