Skip to Content
Merck
All Photos(1)

Key Documents

697761

Sigma-Aldrich

(R,R)-DIPAMP

95%

Synonym(s):

(R,R)-1,2-Bis[(2-methoxyphenyl)(phenylphosphino)]ethane, (R,R)-1,2-Ethanediylbis[(2-methoxyphenyl)phenylphosphine], (R,R)-Ethylenebis[(2-methoxyphenyl)phenylphosphine], [(1R,2R)-(−)-Bis[(2-methoxyphenyl)phenylphosphino]ethane]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3OC6H4P(C6H5)CH2-]2
CAS Number:
Molecular Weight:
458.47
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

optical activity

[α]22/D −81°, c = 1 in chloroform

mp

102-106 °C (lit.)

functional group

phosphine

SMILES string

COc1ccccc1P(CCP(c2ccccc2)c3ccccc3OC)c4ccccc4

InChI

1S/C28H28O2P2/c1-29-25-17-9-11-19-27(25)31(23-13-5-3-6-14-23)21-22-32(24-15-7-4-8-16-24)28-20-12-10-18-26(28)30-2/h3-20H,21-22H2,1-2H3/t31-,32-/m1/s1

InChI key

QKZWXPLBVCKXNQ-ROJLCIKYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R, R)-DIPAMP can be used:
  • As a catalyst in the enantioselective [3+2] cycloaddition of γ-substituted allenoates with β-perfluoroalkyl enones and 3-butynoates with electron-deficient olefins to yield substituted cyclopentenes.
  • To prepare its rhodium metal complexes, which are used as catalysts in asymmetric hydrogenation reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of gas-liquid mass transfer on enantioselectivity in asymmetric hydrogenations
Pestre N, et al.
J. Mol. Catal. A: Chem., 252(1-2), 85-89 (2006)
Phosphine-catalyzed enantioselective [3+2] cycloadditions of γ-substituted allenoates with β-perfluoroalkyl enones
Zhou W, et al.
Chemical Science, 8(6), 4660-4665 (2017)
Highly enantio-, regio-and diastereo-selective one-pot [2+ 3]-cycloaddition reaction via isomerization of 3-butynoates to allenoates
Sampath M and Loh T
Chemical Science, 1(6), 739-742 (2010)
Development of an Asymmetric Hydrogenation Route to (S)-N-Boc-2, 6-Dimethyltyrosine
Praquin C, et al.
Organic Process Research & Development, 15(5), 1124-1129 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service