Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C10H15BrS
CAS Number:
Molecular Weight:
247.20
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12352103
MDL number:
Assay:
97%
Form:
liquid
Quality Segment
assay
97%
form
liquid
refractive index
n20/D 1.529
density
1.240 g/mL at 25 °C
SMILES string
CCCCCCc1ccsc1Br
InChI
1S/C10H15BrS/c1-2-3-4-5-6-9-7-8-12-10(9)11/h7-8H,2-6H2,1H3
InChI key
XQJNXCHDODCAJF-UHFFFAOYSA-N
General description
2-Bromo-3-hexylthiophene is a monomeric precursor that forms bromo terminate polymers. It is synthesized by the bromination of hexylthiophene.
Application
2-Bromo-3-hexylthiophene is majorly used in the formation of π-conjugated conductive polymers (CPs) for the fabrication of organic field effect transistors (OFETs) and organic photovoltaics (OPVs). The Grignard metathesis (GRIM) polymerization of 2-Bromo-3-hexylthiophene terminates a bromine and a proton at both ends to form conductive poly(3-hexylthiophene) (P3HT).
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Chronic 4
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
>230.0 °F
flash_point_c
> 110 °C
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
