Skip to Content
Merck
All Photos(1)

Key Documents

689483

Sigma-Aldrich

1-Ethyl-3-methylimidazolium acetate

97%

Synonym(s):

EMIM Ac

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N2O2
CAS Number:
Molecular Weight:
170.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.5% (HPLC)
97%

form

liquid

impurities

≤5.0% water

refractive index

n20/D 1.501

mp

>30 °C (product can occur as an undercooled melt)

density

1.101 g/cm3 at 20 °C

SMILES string

CC([O-])=O.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.C2H4O2/c1-3-8-5-4-7(2)6-8;1-2(3)4/h4-6H,3H2,1-2H3;1H3,(H,3,4)/q+1;/p-1

InChI key

XIYUIMLQTKODPS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

1-Ethyl-3-methylimidazolium acetate, an aprotic imidazolium room-temperature ionic liquid, is a cellulosic solvent. The pretreatment of sugarcane bagasse with [Emim][Ac] leads to the formation of glucose via enzymatic saccharification.

1-Ethyl-3-methylimidazolium acetate is a catalyst that can be used to perform the acid-free cellulose acetylation process.

Application

1-Ethyl-3-methylimidazolium acetate has been used in the preparation of ionic liquid ferrofluid (ILF).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

327.2 °F - closed cup

Flash Point(C)

164 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proton transfer and polarity changes in ionic liquid?water mixtures: a perspective on hydrogen bonds from ab initio molecular dynamics at the example of 1-ethyl-3-methylimidazolium acetate?water mixtures?Part 1
Brehm, Martin, et al.
Physical Chemistry Chemical Physics, 14.15, 5030-5044 (2012)
Stable and water-tolerant ionic liquid ferrofluids.
Jain N, et al.
ACS Applied Materials & Interfaces, 3(3), 662-667 (2011)
Solvent-free acetylation of cellulose by 1-ethyl-3-methylimidazolium acetate-catalyzed transesterification
Chen, Mingjie and Li, et al.
ACS sustainable chemistry & engineering, 7, 16971-11697 (2019)
Structure and dynamics of 1-ethyl-3-methylimidazolium acetate via molecular dynamics and neutron diffraction
Bowron, D. T., et al.
The Journal of Physical Chemistry B, 114.23, 7760-7768 (2010)
Tobiasz Gabryś et al.
Carbohydrate polymers, 254, 117436-117436 (2020-12-29)
The paper presents the results of a study on the preparation of cellulose-based composite fibres (CEL) with graphene oxide addition (GO). Composite fibres (GO/CEL) were prepared via the wet spinning method from CEL solutions in 1-ethyl-3-methylimidazolium acetate (EMIMAc) that contained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service