Skip to Content
Merck
All Photos(1)

Key Documents

684848

Sigma-Aldrich

Pentacene

sublimed grade, ≥99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14
CAS Number:
Molecular Weight:
278.35
Beilstein:
1912418
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

sublimed grade

Quality Level

Assay

≥99.9% trace metals basis

form

powder

mp

372-374 °C (subl.)

solubility

organic solvents: slightly soluble(lit.)

Orbital energy

HOMO 5 eV 
LUMO 3 eV 

OPV Device Performance

ITO/pentacene/C60/BCP/Al

  • Short-circuit current density (Jsc): 15 mA/cm2
  • Open-circuit voltage (Voc): 0.36 V
  • Fill Factor (FF): 0.5
  • Power Conversion Efficiency (PCE): 2.7 %

semiconductor properties

P-type (mobility=0.4-3 cm2/V·s) (on/off ratio=1E5-1E8)

SMILES string

c1ccc2cc3cc4cc5ccccc5cc4cc3cc2c1

InChI

1S/C22H14/c1-2-6-16-10-20-14-22-12-18-8-4-3-7-17(18)11-21(22)13-19(20)9-15(16)5-1/h1-14H

InChI key

SLIUAWYAILUBJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

TGA/DSC Lot specific traces available upon request

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seonggil Ham et al.
Science advances, 6(28) (2020-09-17)
One-dimensional (1D) devices are becoming the most desirable format for wearable electronic technology because they can be easily woven into electronic (e-) textile(s) with versatile functional units while maintaining their inherent features under mechanical stress. In this study, we designed
Dong Wook Kim et al.
ACS nano, 14(1), 907-918 (2020-01-03)
Transparent flexible transistor array requests large-area fabrication, high integration, high manufacturing throughput, inexpensive process, uniformity in transistor performance, and reproducibility. This study suggests a facile and reliable approach to meet the requirements. We use the Al-coated polymer nanofiber patterns obtained
Efficient thin-film organic solar cells based on pentacene/C60 heterojunctions
Yoo, S.; et al.
Applied Physics Letters, 85, 5427-5427 (2004)
Beomjin Park et al.
ACS nano, 9(8), 8352-8360 (2015-07-07)
Precise graphene patterning is of critical importance for tailor-made and sophisticated two-dimensional nanoelectronic and optical devices. However, graphene-based heterostructures have been grown by delicate multistep chemical vapor deposition methods, limiting preparation of versatile heterostructures. Here, we report one-pot synthesis of
Yuseong Gim et al.
Nanoscale, 7(33), 14100-14108 (2015-08-06)
Atomically-thin molecular layers of aryl-functionalized graphene oxides (GOs) were used to modify the surface characteristics of source-drain electrodes to improve the performances of organic field-effect transistor (OFET) devices. The GOs were functionalized with various aryl diazonium salts, including 4-nitroaniline, 4-fluoroaniline

Articles

Flexible electronic circuits and displays based on organic active materials are future generations of products that may eventually enter mainstream electronics market.

Highly reducing or oxidizing species enhance organic semiconductor conductivity by reducing charge-carrier injection barriers.

Highly reducing or oxidizing species enhance organic semiconductor conductivity by reducing charge-carrier injection barriers.

Highly reducing or oxidizing species enhance organic semiconductor conductivity by reducing charge-carrier injection barriers.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service