Skip to Content
Merck
All Photos(1)

Key Documents

684341

Sigma-Aldrich

(S)-(–)-5-(2-Pyrrolidinyl)-1H-tetrazole

96%

Synonym(s):

(S)-(–)-2-Tetrazol-5-ylpyrrolidine, Proline Tetrazole Catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9N5
CAS Number:
Molecular Weight:
139.16
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

optical activity

[α]20/D -9.0°, c = 1 in methanol

mp

253-258 °C

SMILES string

C1CN[C@@H](C1)c2nnn[nH]2

InChI

1S/C5H9N5/c1-2-4(6-3-1)5-7-9-10-8-5/h4,6H,1-3H2,(H,7,8,9,10)/t4-/m0/s1

InChI key

XUHYQIQIENDJER-BYPYZUCNSA-N

Application

(S)-(−)-5-(2-Pyrrolidinyl)-1H-tetrazole can be used as an organocatalyst:
  • To prepare enantioselective chiral 1,2-oxazines from achiral ketones via an intramolecular Wittig reaction.
  • To synthesize diastereoselective Michael addition products by addition of aliphatic aldehydes to β-nitrostyrene.
  • In the direct asymmetric α-fluorination of linear and branched aldehydes using N-fluorobenzenesulfonamide as the fluorinating agent.

Organocatalyst used for:
  • Direct asymmetric aldol reactions between acetone and aldehydes yielding β -hydroxy ketone and for synthesizing 1,1,1-trichloro-2-alkanols
  • Mannich reactions for synthesis fo α -amino acids and generation of 1,4-diamines
  • Conjugate additions of malonates to enones

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oelke, A. et al.
Synlett, 2548-2548 (2006)
Asymmetric organocatalyzed Michael addition of aldehydes to β-nitrostyrene in ionic liquids
Mevciarova M, et al.
Tetrahedron Asymmetry, 20(20), 2403-2406 (2009)
Sirirat Kumarn et al.
Chemical communications (Cambridge, England), (30), 3211-3213 (2006-10-10)
A sequential, organocatalysed asymmetric reaction to access chiral 1,2-oxazines from achiral ketone starting materials is reported, which proceeds in moderate to good yields and excellent enantioselectivity.
Direct asymmetric α-fluorination of aldehydes
Steiner DD, et al.
Angewandte Chemie (International Edition in English), 117(24), 3772-3776 (2005)
A highly selective, organocatalytic route to chiral 1, 2-oxazines from ketones
Kumarn S, et al.
Chemical Communications (Cambridge, England), 3211-3213 (2006)

Related Content

The Yamamoto group has employed several applications in catalysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service