Skip to Content
Merck
All Photos(1)

Key Documents

682519

Sigma-Aldrich

2,4,6-Trimethylbenzoyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3(C6H2)C(O)Cl
CAS Number:
Molecular Weight:
182.65
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.529

density

1.095 g/mL at 25 °C

functional group

acyl chloride

storage temp.

2-8°C

SMILES string

Cc1cc(C)c(c(C)c1)C(Cl)=O

InChI

1S/C10H11ClO/c1-6-4-7(2)9(10(11)12)8(3)5-6/h4-5H,1-3H3

InChI key

UKRQMDIFLKHCRO-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuanyi Ma et al.
Acta biomaterialia, 95, 319-327 (2018-12-24)
Human induced pluripotent stem cell - derived cardiomyocytes (iPSC-CMs) are regarded as a promising cell source for establishing in-vitro personalized cardiac tissue models and developing therapeutics. However, analyzing cardiac force and drug response using mature human iPSC-CMs in a high-throughput
Rujing Zhang et al.
Lab on a chip, 17(24), 4273-4282 (2017-11-09)
Three-dimensional (3D) in vitro models capturing both the structural and dynamic complexity of the in vivo situation are in great demand as an alternative to animal models. Despite tremendous progress in engineering complex tissue/organ models in the past decade, approaches
Reem Ibrahim Al-Wabli et al.
Chemistry Central journal, 10, 73-73 (2016-12-21)
One of the most popular techniques for cancer detection is the nuclear medicine technique. The present research focuses on Platelet-12-lipoxygenase (P-12-LOX) as a promising target for treating and radio-imaging tumor tissues. Curcumin was reported to inhibit this enzyme via binding
Zhongliang Jiang et al.
Journal of materials chemistry. B, 5(1), 173-180 (2017-01-10)
Cell encapsulation within photopolymerized polyethylene glycol (PEG)-based hydrogel scaffolds has been demonstrated as a robust strategy for cell delivery, tissue engineering, regenerative medicine, and developing in vitro platforms to study cellular behavior and fate. Strategies to achieve spatial and temporal
Cancan Xu et al.
ACS applied materials & interfaces, 10(12), 9969-9979 (2018-02-17)
Cell printing is becoming a common technique to fabricate cellularized printed scaffold for biomedical application. There are still significant challenges in soft tissue bioprinting using hydrogels, which requires live cells inside the hydrogels. Moreover, the resilient mechanical properties from hydrogels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service