Skip to Content
Merck
All Photos(1)

Key Documents

677396

Sigma-Aldrich

Magnesium aluminate, spinel

nanopowder, <50 nm particle size (BET)

Synonym(s):

Magnesium aluminate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO·Al2O3
CAS Number:
Molecular Weight:
142.27
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanopowder

Quality Level

reaction suitability

reagent type: catalyst
core: magnesium

surface area

>30 m2/g

particle size

<50 nm (BET)

mp

2130 °C (lit.)

density

3.64 g/mL at 25 °C (lit.)

SMILES string

[Mg++].[O-][Al]=O.[O-][Al]=O

InChI

1S/2Al.Mg.4O/q;;+2;;;2*-1

InChI key

VPBIQXABTCDMAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • A Review on Processing Polycrystalline Magnesium Aluminate Spinel (MgAl2O4): This review discusses various sintering techniques, material properties, and machinability of polycrystalline magnesium aluminate spinel, highlighting its optical transparency and mechanical properties (Z Shi et al., 2020).
  • Effect of Structure and Composition of Non-Stoichiometry Magnesium Aluminate Spinel on Water Adsorption: This study examines how the structure and composition of magnesium aluminate spinel affect its water adsorption capabilities, focusing on the interaction with gases at different spinel compositions (Y Mordekovitz et al., 2020).
  • Spectroscopic Study of Ordering in Non-Stoichiometric Magnesium Aluminate Spinel: The paper explores the ordering within non-stoichiometric magnesium aluminate spinel and its impact on material properties (V Erukhimovitch et al., 2015).
  • Transparent Polycrystalline Magnesium Aluminate Spinel Fabricated by Spark Plasma Sintering: This research presents the fabrication of transparent polycrystalline magnesium aluminate spinel using spark plasma sintering, analyzing its mechanical and optical properties (M Sokol et al., 2018).
  • A Comparative Study on the Effect of Different Additives on the Formation and Densification of Magnesium Aluminate Spinel: Investigates the effects of various additives on the formation and densification of magnesium aluminate spinel, providing insights into the synthesis and optimization of this ceramic material (SK Mohan et al., 2016).

Legal Information

Product of Engi-Mat Co.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Investigation of Lattice Constant, Sintering, and Properties of Nano Mg-Al Spinels
M.F. Zawrah
Materials Science & Engineering. A, Structural Materials : Properties, Microstructure and Processing, 382, 362-370 (2004)
Microstructural and Mechanical Properties of Nanocrystalline Spinel and Related Composites.
S. Bhaduri, S.B. Bhaduri
Ceramics International, 28, 153-158 (2002)
Sintering Behaviour of MgAl2O4 - A Prospective Anode Material
S. Angappan, L.J. Berchmans, C.O. Augustin
Materials Letters, 58, 2283-2289 (2004)
J C Keller et al.
Dental materials : official publication of the Academy of Dental Materials, 13(1), 62-68 (1997-01-01)
The objective of this work was to examine osteoblast-like cell attachment and morphology in vitro to osteoceramic materials with three different surface morphologies. Osteoceramic composite disks were fabricated from tricalcium phosphate and magnesium-aluminate spinel (MgAl2O4) in a 50 vol% ratio.
Tomohito Kameda et al.
Chemosphere, 73(5), 844-847 (2008-07-25)
Magnesium-aluminum oxide (Mg-Al oxide) obtained by thermal decomposition of Mg-Al layered double hydroxide (Mg-Al LDH) effectively removed HCl from gaseous streams. HCl removal was greater in the presence of added water vapor at all temperatures examined and increased with decreasing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service