Skip to Content
Merck
All Photos(1)

Key Documents

670871

Sigma-Aldrich

(S)-(+)-Pyrrolidine-3-carboxylic acid

≥98.0% (NT)

Synonym(s):

(S)-β-Proline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein:
3537293
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (NT)

form

solid

optical activity

[α]/D 20.0±2.0°, c = 2 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

OC(=O)[C@H]1CCNC1

InChI

1S/C5H9NO2/c7-5(8)4-1-2-6-3-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1

InChI key

JAEIBKXSIXOLOL-BYPYZUCNSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

B R Huck et al.
Organic letters, 1(11), 1717-1720 (2000-06-03)
[formula: see text] Homooligomers of beta-amino acids (S)-3-pyrrolidine-3-carboxylic acid (PCA) and (S)-nipecotic acid (Nip) were studied by circular dichroism (CD) in methanol. In each series, a profound change in the far-UV CD spectrum was observed from monomer to tetramer, but
Alessandra Tolomelli et al.
Amino acids, 41(3), 575-586 (2011-05-05)
The synthesis of unusual cyclic amino acids, that may be envisaged as proline analogs, is an area of great interest for their potential applications as scaffolds for the design of bioactive peptidomimetics or units for the creation of novel foldamers.
Joanna Katarzyńska et al.
Journal of peptide science : an official publication of the European Peptide Society, 14(12), 1283-1294 (2008-09-30)
Immune response suppressors are used in the medical praxis to prevent graft rejection after organ transplantation and in the therapy of some autoimmune diseases. As a continuation of our previous work searching for new, effective suppressors devoid of toxicity, we
Cody Timmons et al.
The Journal of organic chemistry, 70(19), 7634-7639 (2005-09-10)
[reaction: see text] A new halo-Mannich-type reaction is reported using cyclopropyl carbonyl-derived enolates and sulfonyl-protected imines. Chiral oxazolidinones auxiliaries were found to be effective for completely controlling the stereochemistry of the products. Variations in the oxazolidinone, protecting group, and imine
Santi Spampinato et al.
European journal of pharmacology, 469(1-3), 89-95 (2003-06-05)
We previously described a novel endomorphin-1 analogue (Tyr-L-beta-Pro-Trp-Phe-NH(2); Endo1-beta-Pro) more resistant to enzymatic hydrolysis than endomorphin-1 that acts as a mu-opioid receptor agonist. In this study we report that Endo1-beta-Pro, s.c. injected in the mouse, is an effective antinociceptive agent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service