Skip to Content
Merck
All Photos(2)

Key Documents

666610

Sigma-Aldrich

Tetrakis(dimethylamido)hafnium(IV)

packaged for use in deposition systems

Synonym(s):

TDMAH, Tetrakis(dimethylamino)hafnium(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]4Hf
CAS Number:
Molecular Weight:
354.79
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99% (trace metals analysis)

form

low-melting solid

reaction suitability

core: hafnium

mp

26-29 °C (lit.)

density

1.098 g/mL at 25 °C

SMILES string

CN(C)[Hf](N(C)C)(N(C)C)N(C)C

InChI

1S/4C2H6N.Hf/c4*1-3-2;/h4*1-2H3;/q4*-1;+4

InChI key

ZYLGGWPMIDHSEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Alkyl amides of Hafnium provide a convenient and effective atomic layer deposition precursor to smooth and and amorphous hafnium oxide thin films.

Application

Used as precursor for atomic layer deposition of Hafnium Oxide nanolaminates, which are used as a reploacement for Silicon oxide in semiconductor devices.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Savannah ALD System - An Excellent Tool for Atomic Layer Deposition
Monsma D, Becker J
Material Matters, 1(3), 5-5 (2006)
Surface morphology and crystallinity control in the atomic layer deposition (ALD) of hafnium and zirconium oxide thin films
Hausmann DM, Gordon RG
Journal of Crystal Growth, 249, 251-261 (2003)
High Purity Metalorganic Precursors for CPV Device Fabrication
Rushworth S
Material Matters, 5(4) null
Applied Physics Letters, 91, 193503-193503 (2007)

Articles

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service