Skip to Content
Merck
All Photos(3)

Key Documents

665800

Sigma-Aldrich

Polytetrafluoroethylene preparation

60 wt % dispersion in H2O

Synonym(s):

PTFE dispersion

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF2CF2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

concentration

60 wt % dispersion in H2O

SMILES string

FC(=C(F)F)F

InChI

1S/C2F4/c3-1(4)2(5)6

InChI key

BFKJFAAPBSQJPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

PTFE is a synthetic fluropolymer of tetrafluoroethylene. It is a fluorocarbon solid, as it is a high molecular weight compound consisting wholly of carbon and fluorine. PTFE is hydrophobic in nature, it also has the lowest coefficient of friction against any solid.

Application

Owing to its chemical inertness and hydrophobicity, PTFE is widely used in surgical meshes, stents, grafts, and scaffolds for ligament repair. For example, expanded PTFE can be used to prepare vascular grafts.

The invariance of the dielectric constant over a wide frequency range and the high dielectric strength of PTFE makes it a suitable material for cable insulation and high-temperature energy storage application.

It can also be employed as an easy-cleaning, chemically inert, and non-stick coating in food, chemical and pharmaceutical industries.

Features and Benefits

  • High chemical resistivity
  • Low coefficient of friction
  • Flexibility
  • High thermal stability

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

How to Start a Polytetrafluoroethylene (PTFE) Business (Beginners Guide) null
Hocine Bensaadi et al.
The American surgeon, 80(1), 57-65 (2014-01-10)
Funding received from Cousin Biotech, Wervicq Sud, France, and CR Bard Inc., Cranston, RI. The aim of this prospective randomized study was to determine the long-term recurrence and complication rates after small abdominal wall hernia repair with two different bilayer
Aldo Cannata et al.
The Annals of thoracic surgery, 95(5), 1818-1826 (2013-04-10)
Postsurgical intrapericardial adhesions are still considered an unavoidable consequence of cardiothoracic operations. They increase the technical difficulty and the risk of reoperations. The pathogenesis of postsurgical adhesions is a multistep process, and the main key players are (1) loss of
W B Jawaid et al.
The British journal of surgery, 100(13), 1833-1837 (2013-11-15)
The anatomical defect in congenital diaphragmatic hernia (CDH) can usually be closed primarily but prosthetic patch repair may be required in newborns with a deficient diaphragm. High rates of patch failure and hernia recurrence (up to 50 per cent) have
Katherine R Kozak et al.
Lab on a chip, 13(7), 1342-1350 (2013-02-06)
Miniaturization of immunoassays has numerous potential advantages over traditional ELISAs. Here we present a novel approach using patterned planar plates (PPPs). These 'wall-less' plates consist of a 16 × 24 array of 2 mm diameter hydrophilic regions surrounded by a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service