Skip to Content
Merck
All Photos(1)

Key Documents

656631

Sigma-Aldrich

1,3-Bis(2,4,6-trimethylphenyl)imidazolinium chloride

95%

Synonym(s):

1,3-Dimesitylimidazolidinium chloride, 4,5-Dihydro-1,3-bis(2,4,6-trimethylphenyl)-1H-imidazolium chloride, 4,5-Dihydro-1,3-dimesityl-1H-imidazolium chloride, N,N′-(2,4,6-Trimethylphenyl)dihydroimidazolium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27ClN2
CAS Number:
Molecular Weight:
342.91
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

reaction suitability

reagent type: ligand

mp

280-286 °C

SMILES string

[Cl-].Cc1cc(C)c(N2CC[N+](=C2)c3c(C)cc(C)cc3C)c(C)c1

InChI

1S/C21H27N2.ClH/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;/h9-13H,7-8H2,1-6H3;1H/q+1;/p-1

InChI key

COGMCBFILULEOS-UHFFFAOYSA-M

General description

1,3-Bis(2,4,6-trimethylphenyl)imidazolinium chloride is an NHC (N-heterocyclic carbene) ligand which can bind with metal pre-catalysts to form complexes that show high catalytic activity.

Application

N-Heterocyclic Carbene Ligands

Precursor to an N-heterocyclic carbene catalysts used for:
  • A regioselective cycloadditon of terminal acetylenes with azides leading to 1,4-disubstitutedtriazoles. Internal acetylenes can also be used with this catalyst.
  • Markovnikov-type hydration of terminal alkynes
  • Hydrosilylation of ketones and cycloaddition of azides and alkynes
  • Suzuki-Miyaura reactions
  • Luminescence experiments

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Roy and H. Plenio,
Advanced Synthesis & Catalysis, 352, 1014-1022 (2010)
V Poza-Nogueiras et al.
Chemosphere, 199, 68-75 (2018-02-13)
Conventional water treatments are generally inadequate for degradation of emerging pollutants such as ionic liquids (ILs). The use of heterogeneous electro-Fenton (HEF) has attracted great interest, due to its ability to efficiently oxidize a wide range of organic pollutants operating
Silvia Díez-González et al.
Dalton transactions (Cambridge, England : 2003), 39(32), 7595-7606 (2010-07-14)
The preparation of three series of [(NHC)CuX] complexes (NHC = N-heterocyclic carbene, X = Cl, Br, or I) is reported. These syntheses are high yielding and only use readily available starting materials. The prepared complexes were spectroscopically and structurally characterized.
Vonika Ka-Man Au et al.
Journal of the American Chemical Society, 131(25), 9076-9085 (2009-06-03)
A new class of luminescent mononuclear and dinuclear N-heterocyclic carbene-containing gold(III) complexes has been synthesized and characterized. The X-ray crystal structures of most of the complexes have also been determined. Electrochemical studies reveal a ligand-centered reduction originated from the RC--N--CR
Silvia Díez-González et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 12(29), 7558-7564 (2006-09-14)
A versatile and highly efficient catalyst for the Huisgen cycloaddition reaction has been developed. Previously isolated or in situ generated azides yielded 1,2,3-triazoles with differently substituted alkynes in the presence of a [(NHC)CuBr] complex (NHC = N-heterocyclic carbene). Extremely high

Articles

Metal complex-catalyzed cross-coupling reactions of unactivated substrates introduce diverse phosphine ligands in chemical marketplace.

Metal complex-catalyzed cross-coupling reactions of unactivated substrates introduce diverse phosphine ligands in chemical marketplace.

Metal complex-catalyzed cross-coupling reactions of unactivated substrates introduce diverse phosphine ligands in chemical marketplace.

Metal complex-catalyzed cross-coupling reactions of unactivated substrates introduce diverse phosphine ligands in chemical marketplace.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service