Skip to Content
Merck
All Photos(1)

Key Documents

647535

Sigma-Aldrich

Silicon

wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm

Synonym(s):

Silicon element

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si
CAS Number:
Molecular Weight:
28.09
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

crystalline (cubic (a = 5.4037))
wafer (single side polished)

does not contain

dopant

diam. × thickness

3 in. × 0.5 mm

bp

2355 °C (lit.)

mp

1410 °C (lit.)

density

2.33 g/mL at 25 °C (lit.)

semiconductor properties

<100>, N-type

SMILES string

[Si]

InChI

1S/Si

InChI key

XUIMIQQOPSSXEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Silicon wafers are majorly used in electronics as substrate materials. They are also used in a variety of applications like solar cells, microelectronics, and other integrated circuits.[1][2][3]

Physical properties

0 vortex defects. Etch pitch density (EPD) <100 cm-2. Resistivity 1,000- 2,300 Ωcm
Oxygen content: <= 1~1.8 x 1018 /cm3; Carbon content: <= 5 x 1016 /cm3; Boule diameter: 1~8 ″

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

24.7% record efficiency HIT solar cell on thin silicon wafer
Taguchi M, et al.
IEEE journal of Photovoltaics, 4(1), 96-99 (2013)
Stretchable and foldable silicon integrated circuits
Kim D, et al.
Science (New York, N.Y.), 320(5875), 507-511 (2008)
Nanolithography in microelectronics: A review
Seisyan RP
Technical Physics, 56(8), 1061-1061 (2011)
Jaewoo Lee et al.
Journal of nanoscience and nanotechnology, 13(5), 3495-3499 (2013-07-19)
A spin-casting process for fabricating polycrystalline silicon sheets for use as solar cell wafers is proposed, and the parameters that control the sheet thickness are investigated. A numerical study of the fluidity of molten silicon indicates that the formation of
Seungil Park et al.
Journal of nanoscience and nanotechnology, 13(5), 3397-3402 (2013-07-19)
We investigated the thin film growths of hydrogenated silicon by hot-wire chemical vapor deposition with different flow rates of SiH4 and H2 mixture ambient and fabricated thin film solar cells by implementing the intrinsic layers to SiC/Si heterojunction p-i-n structures.

Articles

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

See All

Protocols

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service