Skip to Content
Merck
All Photos(2)

Key Documents

64275

Sigma-Aldrich

Methanesulfonamide

≥97.0% (CHN)

Synonym(s):

Mesylamide, Methanesulfonic amide, Methanesulphonamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2NH2
CAS Number:
Molecular Weight:
95.12
Beilstein:
1740835
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (CHN)

form

solid

mp

85-89 °C (lit.)

SMILES string

CS(N)(=O)=O

InChI

1S/CH5NO2S/c1-5(2,3)4/h1H3,(H2,2,3,4)

InChI key

HNQIVZYLYMDVSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Methanesulfonamide is used in the synthesis of important organic reagents such as N-(2-methylthio-1-p-toluenesufonyl)methanesulfonamide and tert-butyl ((2-(trimethylsilyl)ethyl)sulfonyl)carbamate.
  • It can be used as a source of nitrogen in the conversion of carboxylic acids to corresponding nitriles.
  • It is widely used as a reagent in the synthesis of medicinally important compounds such as derivatives of indole-N-acetamide, methanesulfonamide pyrimidine-substituted 3,5-dihydroxy-6-heptenoates and repertaxin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Convenient Synthesis of Chloronitrobenzonitrile Isomers and Homologs.
Grivsky E M
Bull. Soc. Chim. Belg., 80(3?4), 245-252 (1971)
Development and preliminary optimization of indole-N-acetamide inhibitors of hepatitis C virus NS5B polymerase.
Harper S, et al.
Journal of Medicinal Chemistry, 48(5), 1314-1317 (2005)
Synthesis and biological activity of methanesulfonamide pyrimidine-and N-methanesulfonyl pyrrole-substituted 3, 5-dihydroxy-6-heptenoates, a novel series of HMG-CoA reductase inhibitors.
Watanabe M, et al.
Bioorganic & Medicinal Chemistry, 5(2), 437-444 (1997)
2-Arylpropionic CXC chemokine receptor 1 (CXCR1) ligands as novel noncompetitive CXCL8 inhibitors.
Allegretti M, et al.
Journal of Medicinal Chemistry, 48(13), 4312-4331 (2005)
Sevgi Karakuş et al.
European journal of medicinal chemistry, 44(9), 3591-3595 (2009-03-28)
Due to a continuing effort to develop new antiviral agents, a series of 1-[4-(methanesulfonamido)-3-phenoxyphenyl]-3-alkyl/aryl thioureas 3a-i have been synthesized by the reaction of alkyl/aryl isothiocyanates with 4-amino-2-phenoxymethanesulfonanilide. These derivatives were structurally characterized by the use of spectral techniques and evaluated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service