All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C13H11N3
CAS Number:
Molecular Weight:
209.25
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
32151902
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
solid
mp
211-215 °C (lit.)
SMILES string
Nc1ccccc1-c2nc3ccccc3[nH]2
InChI
1S/C13H11N3/c14-10-6-2-1-5-9(10)13-15-11-7-3-4-8-12(11)16-13/h1-8H,14H2,(H,15,16)
InChI key
YWNXHTNWOQHFRL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Thangaraj Anand et al.
Physical chemistry chemical physics : PCCP, 21(22), 11839-11845 (2019-05-23)
A new optical chemosensor N1 was designed and synthesized by condensing 4-chloro-7-nitrobenzofurazan with 2-aminophenylbenzimidazole. In CH3OH : H2O (1 : 1, v/v) medium, sensor N1 exhibited high selectivity and sensitivity towards Cu2+ and Hg2+ ions by showing a distinct colour change from pale yellow
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service