Skip to Content
Merck
All Photos(1)

Key Documents

636568

Sigma-Aldrich

Acrylamide-2,3,3-d3

98 atom % D, 98% (CP)

Synonym(s):

2-Propenamide-2,3,3-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D2C=CDCONH2
CAS Number:
Molecular Weight:
74.10
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

solid

technique(s)

protein expression: suitable

bp

125 °C/25 mmHg (lit.)

mp

82-86 °C (lit.)

mass shift

M+3

SMILES string

[2H]\C([2H])=C(/[2H])C(N)=O

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)/i1D2,2D

InChI key

HRPVXLWXLXDGHG-FUDHJZNOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mohammad Rizwan Khan et al.
Scientific reports, 7, 41995-41995 (2017-02-06)
The present work describes the outcomes of the assessment on acrylamide contents in a number of thermally treated foods (Arabic coffee Qahwa, coffee and tea) obtained from the Saudi Arabian markets. A total of 56 food samples of different brands
Demetris Kafouris et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 35(10), 1928-1939 (2018-08-28)
The determination of acrylamide in potato products, bakery products and coffee, and the human dietary exposure is reported. The method reported is based on a single extraction step with water, followed by the clean-up of the extract using solid phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service