Skip to Content
Merck
All Photos(1)

Key Documents

635995

Sigma-Aldrich

1,4-Benzodioxane-6-boronic acid

≥95%

Synonym(s):

6-(1,4-Benzodioxanyl)boronic acid, 1,4-Benzodioxan-6-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9BO4
CAS Number:
Molecular Weight:
179.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder

SMILES string

OB(O)c1ccc2OCCOc2c1

InChI

1S/C8H9BO4/c10-9(11)6-1-2-7-8(5-6)13-4-3-12-7/h1-2,5,10-11H,3-4H2

InChI key

SQDUGGGBJXULJR-UHFFFAOYSA-N

Application

Reactant involved in:
  • Addition reactions with 1,8-naphthyridine N-oxides
  • Iodocyclization and palladium-catalyzed coupling reactions for synthesis of pyranoquinolines
  • Suzuki coupling reactions for synthesis of combretastatin analogs
  • Suzuki-Miyaura coupling reactions for synthesis of aryl-substituted oxabenzindoles and methanobenzindoles
  • Palladium-catalyzed oxyarylation of Heck reaction intermediates
  • C-H functionalization of quinones

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service