Skip to Content
Merck
All Photos(1)

Documents

632023

Sigma-Aldrich

3-Hydroxyphenylacetylene

≥95.0%

Synonym(s):

3-Ethynylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6O
CAS Number:
Molecular Weight:
118.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

refractive index

n20/D 1.5840 (lit.)

density

1.083 g/mL at 25 °C (lit.)

SMILES string

Oc1cccc(c1)C#C

InChI

1S/C8H6O/c1-2-7-4-3-5-8(9)6-7/h1,3-6,9H

InChI key

AODMJIOEGCBUQL-UHFFFAOYSA-N

Application

3-Hydroxyphenyl acetylene exhibits chromogenic and fluorogenic properties and hence it can be used as a probe for labeling of bacterial enzymes, which degrade toluene.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

217.4 °F

Flash Point(C)

103 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mary E Kauffman et al.
Journal of microbiological methods, 55(3), 801-805 (2003-11-11)
3-hydroxyphenylacetylene (3-HPA) served as a novel, activity-dependent, fluorogenic and chromogenic probe for bacterial enzymes known to degrade toluene via meta ring fission of the intermediate, 3-methylcatechol. By this direct physiological analysis, cells grown with an aromatic substrate to induce the
"Hydration of aromatic terminal alkynes catalyzed by iron (III) sulfate hydrate under chlorine-free conditions"
Bassetti M, et al.
Tetrahedron Letters, 55(9), 1608-1612 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service