Skip to Content
Merck
All Photos(1)

Key Documents

61718

Sigma-Aldrich

Lavender oil

natural, from Lavandula angustifolia L.

Synonym(s):

Aromaya Raben, Lavandox, Lavandula aetheroleum, Lavandula officinalis oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22
biological source:
Lavandula angustifolia L.

biological source

Lavandula angustifolia L.

Quality Level

quality

natural

refractive index

n20/D 1.46
n20/D 1.461 (lit.)

bp

204 °C (lit.)

density

0.879 g/mL at 25 °C (lit.)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leo B Silenieks et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 20(2), 172-177 (2012-12-05)
Recently, an essential oil of selected quality produced from the flowering tops of Lavandula angustifolia Mill. by steam distillation (Silexan) has been approved in Germany for the treatment of restlessness in case of anxious mood. Based on the observed clinical
Winai Sayorwan et al.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet, 95(4), 598-606 (2012-05-23)
Investigate the effects of lavender oil on the central nervous system, autonomic nervous system, and mood responses in humans after inhalation. Twenty healthy volunteers participated in the experiments. The present study assessed autonomic parameters such as blood pressure, heart rate
P A R Hawken et al.
Physiology & behavior, 105(5), 1117-1123 (2011-12-24)
Growing concerns about the risk of addiction to benzodiazepines have led to increasing interest in alternative therapies to treat anxiety and depression. Lavender oil (Lavendula augustifolia) is reportedly anxiolytic in a number of species but little is known about how
Mei-Yu Huang et al.
The American journal of Chinese medicine, 40(4), 845-859 (2012-07-20)
Lavender essential oil (LEO) is one the most favorite and widely used essential oils in aromatherapy. Many studies have demonstrated its functions in calming, assisting sleep, reducing pain and muscular spasms and its antiseptic function. To date, however, the mechanism
Vikas Kumar
Wiener medizinische Wochenschrift (1946), 163(3-4), 89-94 (2013-01-31)
Silexan is an essential oil produced from Lavandula angustifolia flowers with proven clinical efficacy for the treatment of anxiety disorders. The present study was conducted to assess its anxiolytic activity and to screen for neuropharmacological properties in rats and mice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service