Skip to Content
Merck

616672

DL-Glyceric-2,3,3-d3 acid calcium salt dihydrate

98 atom % D

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
(HOCD2CD(OH)CO2)2Ca•2H2O
CAS Number:
Molecular Weight:
292.28
UNSPSC Code:
12352106
PubChem Substance ID:
MDL number:
Isotopic purity:
98 atom % D
Mass shift:
M+6
Form:
solid


isotopic purity

98 atom % D

form

solid

mass shift

M+6

SMILES string

O.O.[2H]C([2H])(O)C([2H])(O)C(=O)O[Ca]OC(=O)C([2H])(O)C([2H])([2H])O

InChI

1S/2C3H6O4.Ca.2H2O/c2*4-1-2(5)3(6)7;;;/h2*2,4-5H,1H2,(H,6,7);;2*1H2/q;;+2;;/p-2/i2*1D2,2D;;;

InChI key

OKEVQKKVYQHNFW-NJEQVFLESA-L

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library