Skip to Content
Merck
All Photos(1)

Key Documents

61243

Sigma-Aldrich

Copper(II) tartrate hydrate

≥95.0% (calc. on dry substance, RT)

Synonym(s):

Cupric tartrate hydrate, Tartaric acid cupric salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH(OH)CO2]2Cu·xH2O
CAS Number:
Molecular Weight:
211.62 (anhydrous basis)
Beilstein:
4166088
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (calc. on dry substance, RT)

form

solid

reaction suitability

core: copper
reagent type: catalyst

impurities

5-15% water

mp

~275 °C (dec.) (lit.)

SMILES string

O.[Cu++].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

InChI

1S/C4H6O6.Cu.H2O/c5-1(3(7)8)2(6)4(9)10;;/h1-2,5-6H,(H,7,8)(H,9,10);;1H2/q;+2;/p-2/t1-,2-;;/m1../s1

InChI key

MAUZTCHAIPUZJB-OLXYHTOASA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service