Skip to Content
Merck
All Photos(1)

Key Documents

603627

Sigma-Aldrich

1-Pentanol-1-13C

99 atom % 13C, 99% (CP)

Synonym(s):

Amyl alcohol-1-13C, Pentyl alcohol-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)313CH2OH
CAS Number:
Molecular Weight:
89.14
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

bp

136-138 °C (lit.)

mp

-78 °C (lit.)

density

0.820 g/mL at 25 °C

mass shift

M+1

SMILES string

CCCC[13CH2]O

InChI

1S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3/i5+1

InChI key

AMQJEAYHLZJPGS-HOSYLAQJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Whasley Ferreira Duarte et al.
Journal of food science, 76(9), C1307-C1318 (2012-03-16)
The effect of Lactobacillus fermentum in co-inoculation with Saccharomyces cerevisiae UFLA CA11 on the quality of cachaça (sugar cane spirit) was evaluated. The co-inoculation was first evaluated in flask fermentation, and subsequently, yeast and bacteria were co-cultured at approximately 10(5)
Acid/base- and anion-controllable organogels formed from a urea-based molecular switch.
Sheng-Yao Hsueh et al.
Angewandte Chemie (International ed. in English), 49(48), 9170-9173 (2010-10-19)
S Wellert et al.
Journal of colloid and interface science, 325(1), 250-258 (2008-06-24)
Most toxic industrial chemicals and chemical warfare agents are hydrophobic and can only be solubilized in organic solvents. However, most reagents employed for the degradation of these toxic compounds can only be dissolved in water. Hence, microemulsions are auspicious media
Kristine von Maltzan et al.
Alcohol (Fayetteville, N.Y.), 45(1), 1-9 (2010-09-17)
Enzyme-linked immunosorbent assays (ELISAs) are frequently used in studies on cytokine production in response to treatment of cell cultures or laboratory animals. When an ELISA assay is performed on cell culture supernatants, samples often contain the treatment agents. The purpose
Nadav Amdursky et al.
The journal of physical chemistry. A, 115(24), 6481-6487 (2011-05-19)
Time-resolved emission techniques were employed to study the nonradiative process of thioflavin-T (ThT) in 1-propanol, 1-butanol, and 1-pentanol as a function of the hydrostatic pressure. Elevated hydrostatic pressure increases the alcohol viscosity, which in turn strongly influences the nonradiative rate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service