All Photos(3)
About This Item
Linear Formula:
6LiF
CAS Number:
Molecular Weight:
25.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
isotopic purity
95 atom % 6Li
Quality Level
Assay
99% (CP)
form
solid
mp
842 °C
mass shift
M-1
storage temp.
room temp
SMILES string
[6Li+].[F-]
InChI
1S/FH.Li/h1H;/q;+1/p-1/i;1-1
InChI key
PQXKHYXIUOZZFA-HCMAANCNSA-M
Looking for similar products? Visit Product Comparison Guide
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Lact. - Repr. 2
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Paola Francisca Caprile et al.
Health physics, 104(2), 218-223 (2013-01-01)
The characteristics of the thermoluminescence glow curve and dosimetric peak of LiF:Mg,Ti (TLD-100) crystals are studied, with emphasis on the evaluation of the influence of the irradiation dose and heating rate on the dosimetric peak (peak 5) trapping parameters. These
Christian Bernhardsson et al.
Health physics, 103(6), 740-749 (2012-11-01)
Laboratory studies have shown that ordinary household salt (NaCl) exhibits several promising properties for retrospective dosimetry; e.g., a linear dose response and a low detection limit, down to a few mGy or even less. In an attempt to test NaCl
Asa Carlsson Tedgren et al.
Medical physics, 38(10), 5539-5550 (2011-10-14)
High energy photon beams are used in calibrating dosimeters for use in brachytherapy since absorbed dose to water can be determined accurately and with traceability to primary standards in such beams, using calibrated ion chambers and standard dosimetry protocols. For
Chilin Li et al.
Nano letters, 12(3), 1241-1246 (2012-02-07)
The thermodynamically required redistribution of ions at given interfaces is being paid increased attention. The present investigation of the contact LiF/TiO(2) offers a highly worthwhile example, as the redistribution processes can be predicted and verified. It consists in Li ion
Basile F E Curchod et al.
Chimia, 67(4), 218-221 (2013-08-24)
The implementation of local control theory using nonadiabatic molecular dynamics within the framework of linear-response time-dependent density functional theory is discussed. The method is applied to study the photoexcitation of lithium fluoride, for which we demonstrate that this approach can
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service