Skip to Content
Merck
All Photos(3)

Key Documents

595314

Sigma-Aldrich

1-Methylpyrazole-4-boronic acid pinacol ester

95%

Synonym(s):

1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-Methyl-4-pyrazoleboronic acid pinacol ester, 2-(1-Methylpyrazol-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-methylpyrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17BN2O2
CAS Number:
Molecular Weight:
208.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

59-64 °C (lit.)

SMILES string

Cn1cc(cn1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C10H17BN2O2/c1-9(2)10(3,4)15-11(14-9)8-6-12-13(5)7-8/h6-7H,1-5H3

InChI key

UCNGGGYMLHAMJG-UHFFFAOYSA-N

Application

Reagent used for
  • Suzuki-Miyaura cross-coupling reactions
  • Transesterification reactions

Reagent used for preparation of
  • Aminothiazoles as γ-secretase modulators
  • Amino-pyrido-indol-carboxamides, as potential JAK2 inhibitors for myeloproliferative disorders therapy
  • Pyridine derivatives as TGF-β1 and activin A signalling inhibitors
  • MK-2461 analogs as inhibitors of c-Met kinase for the treatment of cancer
Stable alternative to the boronic acid for Suzuki-Miyaura palladium-catalyzed cross-coupling

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason D Katz et al.
Journal of medicinal chemistry, 54(12), 4092-4108 (2011-05-26)
c-Met is a transmembrane tyrosine kinase that mediates activation of several signaling pathways implicated in aggressive cancer phenotypes. In recent years, research into this area has highlighted c-Met as an attractive cancer drug target, triggering a number of approaches to
Thomas Lübbers et al.
Bioorganic & medicinal chemistry letters, 21(21), 6554-6558 (2011-09-20)
We herein report the discovery of a new γ-secretase modulator class with an aminothiazole core starting from a HTS hit (3). Synthesis and SAR of this series are discussed. These novel compounds demonstrate moderate to good in vitro potency in
Suzuki-Miyaura cross-coupling reaction of aryl and heteroaryl pinacol boronates for the synthesis of 2-substituted pyrimidines
Asano, S.; Kamioka, S.; Isobe, Y.
Tetrahedron, 68, 272-279 (2012)
Guopin Xu et al.
Organic letters, 7(21), 4605-4608 (2005-10-08)
[reaction: see text] The synthesis of a series of NHC building blocks that can then be incorporated into more complicated structures by palladium catalysis is reported. This approach is used for the synthesis of three amino acids containing NHC side
Direct conversion of pinacol arylboronic esters to aryl triolborates
Li, G.-Q.; et al.
Chemistry Letters (Jpn), 40, 702-704 (2011)

Articles

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service