Skip to Content
Merck
All Photos(2)

Key Documents

594598

Sigma-Aldrich

2-Bromo-2′,4′-dihydroxyacetophenone

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7BrO3
CAS Number:
Molecular Weight:
231.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

128-132 °C (lit.)

SMILES string

Oc1ccc(c(O)c1)C(=O)CBr

InChI

1S/C8H7BrO3/c9-4-8(12)6-2-1-5(10)3-7(6)11/h1-3,10-11H,4H2

InChI key

RAULLGKGLGXMOM-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R K Munns et al.
Journal of chromatography, 442, 209-218 (1988-06-17)
A method is presented for the derivatization and determination of carboxylic acids by high-performance liquid chromatography with electrochemical and ultraviolet detection. The derivatizing reagents used in this study were synthesized, and their suitability was investigated for determination of drugs and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service