Skip to Content
Merck
All Photos(2)

Key Documents

587583

Sigma-Aldrich

Zinc phosphate

99.998% trace metals basis

Synonym(s):

Pigment White 32, Trizinc diphosphate, Zinc orthophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn3(PO4)2
CAS Number:
Molecular Weight:
386.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder and chunks
solubility:
H2O: insoluble(lit.)

grade

for analytical purposes

Quality Level

Assay

99.998% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: zinc

impurities

≤25.0 ppm Trace Metal Analysis

mp

900 °C (lit.)

solubility

H2O: insoluble(lit.)

density

4.0 g/mL (lit.)

SMILES string

[Zn++].[Zn++].[Zn++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/2H3O4P.3Zn/c2*1-5(2,3)4;;;/h2*(H3,1,2,3,4);;;/q;;3*+2/p-6

InChI key

LRXTYHSAJDENHV-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mahtab Memarpour et al.
Pediatric dentistry, 33(7), 501-504 (2012-02-23)
This study's purpose was to compare the ability of 5 luting cements to reduce microleakage at stainless steel crown (SSC) margins on primary molar teeth. Standard preparations were performed on 100 extracted primary molar teeth for SSC restoration. After fitting
R Hmaidouch et al.
International journal of computerized dentistry, 14(3), 219-226 (2011-12-07)
To evaluate the influence of two different tooth preparation forms, two luting space settings and two cement types on the marginal and internal adaptation of all-ceramic crown copings produced using Cerec3 CAD/CAM system. Forty working stone dies were made from
Masayuki Takimoto et al.
Journal of dentistry, 40(2), 131-138 (2011-12-03)
The surface free energy and dentine bond strength of self-adhesive cements were examined after the removal of temporary cements. The labial dentine surfaces of bovine mandibular incisors were wet ground with #600-grit SiC paper. Acrylic resin blocks were luted to
Cui Li et al.
Annals of botany, 123(1), 57-68 (2018-07-19)
The pathways whereby foliar-applied nutrients move across the leaf surface remain unclear. The aim of the present study was to examine the pathways by which foliar-applied Zn moves across the sunflower (Helianthus annuus) leaf surface, considering the potential importance of
Neilor Mateus Antunes Braga et al.
Brazilian dental journal, 23(1), 49-53 (2012-03-31)
This in vitro study compared different ultrasonic vibration modes for intraradicular cast post removal. The crowns of 24 maxillary canines were removed, the roots were embedded in acrylic resin blocks, and the canals were treated endodontically. The post holes were

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service