Skip to Content
Merck

Skip To

577952

Fmoc-Asp-OtBu-15N

98 atom % 15N

Synonym(s):

L-Aspartic-15N acid, N-Fmoc, 1−tert-butyl ester

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
HO2CCH2CH(15NH-Fmoc)CO2C(CH3)3
Molecular Weight:
412.44
PubChem Substance ID:
UNSPSC Code:
12352209
Isotopic purity:
98 atom % 15N
Mass shift:
M+1
Form:
solid
Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service


isotopic purity

98 atom % 15N

form

solid

technique(s)

bio NMR: suitable

application(s)

peptide synthesis

functional group

Fmoc

mass shift

M+1

SMILES string

OC(C[C@H]([15NH]C(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)C(CC(C)(C)C)=O)=O

InChI

1S/C24H27NO5/c1-24(2,3)13-21(26)20(12-22(27)28)25-23(29)30-14-19-17-10-6-4-8-15(17)16-9-5-7-11-18(16)19/h4-11,19-20H,12-14H2,1-3H3,(H,25,29)(H,27,28)/t20-/m0/s1/i25+1

InChI key

PUNKRZNOUPSOSL-HWAMHAJJSA-N

General description

Fmoc-Asp-OtBu-15N 98 atom % 15N is a high-purity isotope product with a unique chemical profile that belongs to the class of isotopes. Produced by Sigma-Aldrich, we are committed to providing quality isotope products and a reliable supply chain. Available in industrial and pre-pack quantities.

Application

- Protein structure determination by NMR spectroscopy
- Nitrogen fixation research in agricultural science
- Isotope ratio Mass spectrometry applications
- Mass spectrometry calibration

Features and Benefits

Features
  • Cost-effective solution for Fmoc-Asp-OtBu-15N 98 atom % 15N applications.
  • Versatile use in different formulations involving Fmoc-Asp-OtBu-15N 98 atom % 15N.
  • Exceptional stability and compatibility with various ingredients.
  • High purity of Fmoc-Asp-OtBu-15N 98 atom % 15N ensures optimal performance.


Benefits
  • Reduces time to market for products utilizing Fmoc-Asp-OtBu-15N 98 atom % 15N.
  • Improves overall product quality and consumer satisfaction.
  • Increases market competitiveness of products containing Fmoc-Asp-OtBu-15N 98 atom % 15N.
  • Enhances product efficacy and reliability in formulations.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact
Stable Isotopes Customer Service.


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
594075588628605263
description

98 atom % 15N

description

98 atom % 15N

description

99 atom % 13C

description

98 atom % 13C, 99% (CP)

mass shift

M+1

mass shift

M+1

mass shift

M+1

mass shift

M+1

technique(s)

bio NMR: suitable

technique(s)

bio NMR: suitable

technique(s)

bio NMR: suitable

technique(s)

bio NMR: suitable

isotopic purity

98 atom % 15N

isotopic purity

98 atom % 15N

isotopic purity

99 atom % 13C

isotopic purity

98 atom % 13C

form

solid

form

solid

form

solid

form

solid

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

functional group

Fmoc

functional group

Fmoc

functional group

Fmoc


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters