Skip to Content
Merck
All Photos(1)

Documents

561479

Sigma-Aldrich

4-Aminopiperidine

Synonym(s):

4-Piperidinamine, Piperidin-4-ylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H10NNH2
CAS Number:
Molecular Weight:
100.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

refractive index

n20/D 1.4910 (lit.)

Quality Level

density

0.945 g/mL at 25 °C (lit.)

SMILES string

NC1CCNCC1

InChI

1S/C5H12N2/c6-5-1-3-7-4-2-5/h5,7H,1-4,6H2

InChI key

BCIIMDOZSUCSEN-UHFFFAOYSA-N

Gene Information

Application

Reactant for synthesis of:
  • Cdk5/p25 kinase inhibitors
  • Antimalarials
  • Selective cyclin-dependent kinase 4/6 inhibitors
  • IKKβ inhibitors
  • Orally bioavailable P2Y12 antagonists for inhibition of platelet aggregation
  • Human and murine soluble epoxide hydrolase inhibitors

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.1 °F - closed cup

Flash Point(C)

45.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service