Skip to Content
Merck
All Photos(1)

Key Documents

559105

Sigma-Aldrich

Ethyl 2-nitrobenzoylacetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4COCH2CO2CH2CH3
CAS Number:
Molecular Weight:
237.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

30-34 °C (lit.)

functional group

ester
ketone
nitro

SMILES string

CCOC(=O)CC(=O)c1ccccc1[N+]([O-])=O

InChI

1S/C11H11NO5/c1-2-17-11(14)7-10(13)8-5-3-4-6-9(8)12(15)16/h3-6H,2,7H2,1H3

InChI key

OWZNCVIBJQPNEF-UHFFFAOYSA-N

Application

Ethyl 2-nitrobenzoylacetate may be used to synthesize ethyl 2-bromo-2′-nitrobenzoylacetate and 4-hydroxy-2(1H)-quinolone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

4-Hydroxy-2 (1H)-quinolone. Part 1: Synthesis and Reactions.
Abdou MM.
Arabian Journal of Chemistry (2014)
Thiazolo [3, 4-b] indazole, a ring-fused tetravalent sulfur thiazole system.
Potts KT and Marshall JL.
The Journal of Organic Chemistry, 41(1), 129-133 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service