Skip to Content
Merck
All Photos(3)

Key Documents

554847

Sigma-Aldrich

Barium oxide

99.99% trace metals basis

Synonym(s):

Baria, Barium monoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaO
CAS Number:
Molecular Weight:
153.33
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: barium

density

5.72 g/mL at 25 °C (lit.)

SMILES string

O=[Ba]

InChI

1S/Ba.O

InChI key

QVQLCTNNEUAWMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Barium Oxide (BaO) is a white ionic crystal that has a non-flammable nature. It is a cubic structured compound that can be prepared by different methodologies, such as thermal decomposition of barium nitrate, and by heating barium carbonate with carbon black or tar.

Application

BaO can be used in a variety of applications such as:
  • network modifier in the development of cathode ray tubes
  • fabrication of glasses such as crown and borosilicate glasses
  • storage of nitrogen gas
  • preparation of biodiesel for internal combustion engines

Features and Benefits

Used in the synthesis of the high-κ material BST (barium strontium titanate). Used in the study of NOX storage.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The characterization of waste cathode-ray tube glass
Mear F, et al.
Waste Management (New York, N.Y.), 26(12), 1468-1476 (2006)
Emission analysis of Azolla methyl ester with BaO nano additives for IC engine
Kannan D and Christraj W
Energy Sources, Part A: Recovery, Utilization, and Environmental Effects, 40(10), 1234-1241 (2018)
Accommodation of excess oxygen in group II monoxides
Middleburgh SC, et al.
Journal of the American Ceramic Society. American Ceramic Society, 96(1), 308-311 (2013)
Barium oxide
Handbook of Inorganic Chemicals, 529, 77-94 (2003)
Modification in structural and optical properties of CeO2 doped BaO-B2O3 glasses
Singh GP and Singh DP
Journal of Molecular Structure, 1012(12), 137-140 (2012)

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service