Skip to Content
Merck
All Photos(1)

Key Documents

547190

Sigma-Aldrich

Berbamine dihydrochloride

Synonym(s):

berbenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H40N2O6 ·2HCl
CAS Number:
Molecular Weight:
681.65
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

optical activity

[α]20/D +116°, c = 1% in ethanol

mp

250-253 °C (lit.)

SMILES string

Cl.Cl.COc1cc2CCN(C)[C@H]3Cc4ccc(Oc5cc(C[C@H]6N(C)CCc7cc(OC)c(OC)c(Oc1cc23)c67)ccc5O)cc4

InChI

1S/C37H40N2O6.2ClH/c1-38-14-12-24-19-32(41-3)33-21-27(24)28(38)16-22-6-9-26(10-7-22)44-31-18-23(8-11-30(31)40)17-29-35-25(13-15-39(29)2)20-34(42-4)36(43-5)37(35)45-33;;/h6-11,18-21,28-29,40H,12-17H2,1-5H3;2*1H/t28-,29+;;/m0../s1

InChI key

USRXDYNDPPUBSG-KKXMJGKMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of and anti-leukemia active berbamine derivatives

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cai-Mei Zhang et al.
Journal of cardiovascular pharmacology, 58(1), 40-48 (2011-07-15)
Berbamine (BM), a natural compound derived from Berberis vulgaris L, has been reported to inhibit cardiac contractile function at higher concentrations. Here, we report that BM had concentration-dependent biphasic effects on myocardial contraction in Langendorff-perfused rat hearts, that is, at
Rong Liu et al.
Journal of pharmaceutical sciences, 99(7), 3266-3275 (2010-01-30)
Multidrug resistance (MDR) mediated by the overexpression of the drug efflux protein P-glycoprotein is one of the major obstacles to successful cancer chemotherapy. The development of safe and effective MDR-reversing agents is an important approach to addressing this problem clinically.
Yun Liang et al.
Journal of Zhejiang University. Science. B, 12(7), 568-574 (2011-07-05)
The cytotoxic effect of berbamine on chronic myeloid leukemia (CML) cell line KU812 was evaluated, and the mechanisms of its action were explored. The effect of berbamine on the KU812 cell growth was determined by methyl thiazolyl tetrazolium (MTT) assay.
Zhaofu Wang et al.
Journal of mass spectrometry : JMS, 43(3), 327-335 (2007-10-31)
Electrospray ionization mass spectrometry (ESI-MS) was used to investigate the binding of 13 alkaloids to two GC-rich DNA duplexes which are critical sequences in human survivin promoter. Negative ion ESI-MS was first applied to screen the binding of the alkaloids
Yanli Cui et al.
European journal of medicinal chemistry, 54, 867-872 (2012-05-29)
A series of berbamine glycosides was designed, synthesized and evaluated as a new class of antitumor agents. An efficient glycosylation route was developed for berbamide derivatives. The newly synthesized glycosides were evaluated for their cytotoxic activity in vitro against a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service